EMAIL THIS PAGE TO A FRIEND
Circulation research
Deficiency of the sialyltransferase St3Gal4 reduces Ccl5-mediated myeloid cell recruitment and arrest: short communication.
PMID 24425712
Abstract
Sialylation by α2,3-sialyltransferases has been shown to be a crucial glycosylation step in the generation of functional selectin ligands. Recent evidence suggests that sialylation also affects the binding of chemokines to their corresponding receptor. Because the chemokine receptors for Ccl5 and Ccl2 are important in atherogenic recruitment of neutrophils and monocytes, we here investigated the role of α2,3-sialyltransferase IV (ST3Gal-IV) in Ccl5- and Ccl2-mediated myeloid cell arrest and further studied its relevance in a mouse model of atherosclerosis. St3Gal4-deficient myeloid cells showed a reduced binding of Ccl5 and an impaired Ccl5-triggered integrin activation. Correspondingly, Ccl5-induced arrest on tumor necrosis factor-α-stimulated endothelium was almost completely abrogated, as observed in flow chamber adhesion assays and during ex vivo perfusion or intravital microscopy of carotid arteries. Moreover, Ccl5-triggered neutrophil and monocyte extravasation into the peritoneal cavity was severely reduced in St3Gal4(-/-) mice. In contrast, St3Gal4 deficiency did not significantly affect Ccl2 binding and only marginally decreased Ccl2-induced flow arrest of myeloid cells. In agreement with the crucial role of leukocyte accumulation in atherogenesis, and the importance of Ccl5 chemokine receptors mediating myeloid cell recruitment to atherosclerotic vessels, St3Gal4 deficiency drastically reduced the size, stage, and inflammatory cell content of atherosclerotic lesions in Apoe(-/-) mice on high-fat diet. In summary, these findings identify ST3Gal-IV as a promising target to reduce inflammatory leukocyte recruitment and arrest.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N7271 α(2→3) Neuraminidase from Streptococcus pneumoniae, buffered aqueous solution
N5521 α(2→3,6) Neuraminidase from Clostridium perfringens (C. welchii), recombinant, expressed in E. coli, buffered aqueous solution, ≥250 units/mg protein
N8271 α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, recombinant, expressed in E. coli, buffered aqueous solution
N3786
α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, Proteomics Grade, suitable for MALDI-TOF MS
S1951
α-2,3-Sialyltransferase from Pasteurella multocida, recombinant, expressed in E. coli BL21, ≥2 units/mg protein
N5254 Neuraminidase Agarose from Clostridium perfringens (C. welchii), Type VI-A, ammonium sulfate suspension
N2876
Neuraminidase from Clostridium perfringens (C. welchii), Suitable for manufacturing of diagnostic kits and reagents, Type V, lyophilized powder
N3001
Neuraminidase from Clostridium perfringens (C. welchii), Type VI, lyophilized powder, 6-10 units/mg protein (using 4MU-NANA), 2-5 units/mg protein (mucin)
N5631
Neuraminidase from Clostridium perfringens (C. welchii), Type VIII, lyophilized powder, 10-20 units/mg protein (using 4MU-NANA), 3.5-8.0 units/mg protein (mucin)
N2133
Neuraminidase from Clostridium perfringens (C. welchii), Type X, lyophilized powder, ≥50 units/mg protein (using 4MU-NANA)
N7885
Neuraminidase from Vibrio cholerae, Type III, buffered aqueous solution, 0.2 μm filtered, 1-5 units/mg protein (Lowry, using NAN-lactose)
N6514
Neuraminidase from Vibrio cholerae, Type II, buffered aqueous solution, 8-24 units/mg protein (Lowry, using NAN-lactose)