EMAIL THIS PAGE TO A FRIEND
Kidney international
Impaired vitamin K recycling in uremia is rescued by vitamin K supplementation.
PMID 24429407
Abstract
In chronic kidney disease, vitamin K-dependent proteins, including the calcification inhibitor matrix Gla protein, are largely uncarboxylated indicating that functional vitamin K deficiency may contribute to uremic vascular calcification. Since the effects of uremia on the vitamin K cycle are unknown, we investigated the influence of uremia and vitamin K supplementation on the activity of the vitamin K cycle and extraosseous calcification. Uremia was induced in rats by an adenine-supplemented diet and vitamin K1 or K2 was administered over 4 and 7 weeks. After 4 weeks of adenine diet, the activity of the vitamin K cycle enzyme γ-carboxylase but not the activities of DT-diaphorase or vitamin K epoxide reductase were reduced. Serum levels of undercarboxylated matrix Gla protein increased, indicating functional vitamin K deficiency. There was no light microscopy-detectable calcification at this stage but chemically determined aortic and renal calcium content was increased. Vitamin K treatment reduced aortic and renal calcium content after 4 weeks. Seven weeks of uremia induced overt calcification in the aorta, heart, and kidneys; however, addition of vitamin K restored intrarenal γ-carboxylase activity and overstimulated it in the liver along with reducing heart and kidney calcification. Thus, uremic vitamin K deficiency may partially result from a reduction of the γ-carboxylase activity which possibly contributes to calcification. Pharmacological vitamin K supplementation restored the vitamin K cycle and slowed development of soft tissue calcification in experimental uremia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1570428
2-Propanol, United States Pharmacopeia (USP) Reference Standard
C3H8O
PHR1072
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
C3H8O
91237
2-Propanol, analytical standard
C3H8O
M1390
3,3′-Methylene-bis(4-hydroxycoumarin)
C19H12O6
442418
4-Methylphenol, analytical standard
C7H8O
42429
p-Cresol, analytical standard
C7H8O
W233706
p-Cresol, ≥99%, FG
C7H8O
47773
Phylloquinone (K1), analytical standard
C31H46O2
P1609000
Phytomenadione, European Pharmacopoeia (EP) Reference Standard
C31H46O2
1538006
Phytonadione, United States Pharmacopeia (USP) Reference Standard
C31H46O2
PHR1078
Phytonadione, Pharmaceutical Secondary Standard; Certified Reference Material
C31H46O2
S8157
Silver nitrate, BioXtra, >99% (titration)
AgNO3
S7276
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
AgNO3
204390
Silver nitrate, 99.9999% trace metals basis
AgNO3
85229
Silver nitrate, tested according to Ph.Eur.
AgNO3
792276
Silver nitrate, anhydrous, ≥99.999% trace metals basis
AgNO3
248762
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh
AgNO3
85193
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
AgNO3
11691
Silver standard for ICP, for ICP, ready-to-use, in nitric acid
AgNO3
330361
Soda ash, (99% as sodium carbonate or 58% as sodium oxide), technical grade, light
CNa2O3
791768
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
CNa2O3
S7795
Sodium carbonate, BioXtra, ≥99.0%
CNa2O3
451614
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
CNa2O3
71345
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
CNa2O3
71363
Sodium carbonate, certified reference material for titrimetry, certified by BAM, according to ISO 17025, ≥99.5%
CNa2O3
28-2170
Sodium carbonate, JIS special grade, ≥99.5%
CNa2O3
72784
Sodium carbonate concentrate, Na2CO3 72 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 7
CNa2O3
56169
Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC
CNa2O3
490741
Sodium carbonate-12C, 99.9 atom % 12C
12CNa2O3
43617
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
HNaO