EMAIL THIS PAGE TO A FRIEND
Arthritis & rheumatology (Hoboken, N.J.)
Fas signaling in macrophages promotes chronicity in K/BxN serum-induced arthritis.
PMID 24431281
Abstract
A nonapoptotic role of Fas signaling has been implicated in the regulation of inflammation and innate immunity. This study was undertaken to elucidate the contribution of Fas signaling in macrophages to the development of arthritis. K/BxN serum-transfer arthritis was induced in a mouse line in which Fas was conditionally deleted in the myeloid lineage (Cre(LysM) Fas(flox/flox) mice). The arthritis was assessed clinically and histologically. Expression of interleukin-1β (IL-1β), CXCL5, IL-10, IL-6, and gp96 was determined by enzyme-linked immunosorbent assay. Bone marrow-derived macrophages were activated with IL-1β and gp96. Cell phenotype and apoptosis were analyzed by flow cytometry. Arthritis onset in Cre(LysM) Fas(flox/flox) mice was comparable with that observed in control mice; however, resolution was accelerated during the chronic phase. The attenuated arthritis was associated with reduced articular expression of the endogenous Toll-like receptor 2 (TLR-2) ligand gp96 and the neutrophil chemotactic chemokine CXCL5, and enhanced expression of IL-10. Activation with IL-1β or gp96 induced increased IL-10 expression in Fas-deficient murine macrophages compared with control macrophages. IL-10 suppressed IL-6 and CXCL5 expression induced by IL-1β plus gp96. IL-1β-mediated activation of ERK, which regulates IL-10 expression, was increased in Fas-deficient mouse macrophages. Taken together, our findings indicate that impaired Fas signaling results in enhanced expression of antiinflammatory IL-10 and reduced expression of gp96, and these effects are associated with accelerated resolution of inflammation during the chronic phase of arthritis. These observations suggest that strategies to reduce endogenous TLR ligands and increase IL-10 may be beneficial in the treatment of rheumatoid arthritis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4500797
Anti-AKT antibody produced in rabbit, affinity isolated antibody
SAB4500799
Anti-AKT antibody produced in rabbit, affinity isolated antibody
SAB4500800
Anti-AKT antibody produced in rabbit, affinity isolated antibody
SAB4500802
Anti-AKT antibody produced in rabbit, affinity isolated antibody
SAB4300335
Anti-AKT1 (Ab-308) antibody produced in rabbit, affinity isolated antibody
SAB4300575
Anti-AKT1 (Ab-450) antibody produced in rabbit, affinity isolated antibody
SAB4300334
Anti-AKT1 (Ab-473) antibody produced in rabbit, affinity isolated antibody
GW22757 Anti-AKT1 (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB3500216
Anti-Akt1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4500007
Anti-AKT1 antibody produced in rabbit, affinity isolated antibody
AV06008 Anti-AKT1 antibody produced in rabbit, affinity isolated antibody
HPA002891
Anti-AKT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SRP3083 IL-1 beta human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
SRP6169
IL-1β human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)
SAB1400005
Monoclonal Anti-AKT1 antibody produced in mouse, clone 6D6, purified immunoglobulin, buffered aqueous solution
SAB1400007
Monoclonal Anti-AKT1 antibody produced in mouse, clone 2H1, purified immunoglobulin, buffered aqueous solution
SAB1400008
Monoclonal Anti-AKT1 antibody produced in mouse, clone 4B9, purified immunoglobulin, buffered aqueous solution
SAB1400006
Monoclonal Anti-AKT1 antibody produced in mouse, clone 6G6, purified immunoglobulin, buffered aqueous solution
WH0000207M3
Monoclonal Anti-AKT1 antibody produced in mouse, clone 2E11, purified immunoglobulin, buffered aqueous solution
R2650 Monoclonal Anti-Rac antibody produced in mouse, clone 23A8, purified immunoglobulin, buffered aqueous solution
PLA0238
Rabbit anti-AKT1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
SAB2501934
Anti-AKT1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB4500803
Anti-AKT antibody produced in rabbit, affinity isolated antibody