EMAIL THIS PAGE TO A FRIEND
Saudi journal of kidney diseases and transplantation : an official publication of the Saudi Center for Organ Transplantation, Saudi Arabia
Metabolic status of 1088 patients after renal transplantation: assessment of twelve years monitoring in Algiers Mustapha Hospital.
PMID 24434409
Abstract
Since the introduction of monitoring levels of immunosuppressive medications in our service in July 2000, 1088 kidney transplant patients were received for therapeutic drug monitoring and regular follow-up. The aim of this study was to retrospectively analyze the data on these renal graft patients in Algeria and correlate with our 12 years' experience with calcineurin inhibitor (CNI) measurements. In addition, during this period, we also examined other bioche-mical parameters. The analysis was focused on the difference of effect of cyclosporin A (CsA; 623 patients) and Tacrolimus (Tac; 465 patients) on lipid and glucose metabolism and their side-effects, if any, on the renal function. The mean age at the time of transplantation was 36.1 years. A great majority of the transplanted kidneys had been taken from living related donors (88.6%). Three-quarters of all grafts were transplanted in our country (79.5%). Dyslipidemia and renal dysfunction were the most common adverse effects of CsA and Tac exposure, with a frequency of 21.4% and 10.3%, respectively. Both the CNIs had a similar effect on the lipid levels. The highest incidence occurred at 3-12 months after renal graft. Tac seemed to have more side-effects on glycemia, causing the onset of diabetes mellitus more than two-fold than CsA (6.9% vs. 3.1%). A significant difference was observed during 12-24 months after transplantation. However, Tac was associated with the most favorable effects on renal function estimated with the Modification of Diet in Renal Disease (MDRD) formula.