EMAIL THIS PAGE TO A FRIEND
Clinical neuropharmacology
Reduction in the free radical status and clinical benefit of repeated intrathecal triamcinolone acetonide application in patients with progressive multiple sclerosis.
PMID 24434528