EMAIL THIS PAGE TO A FRIEND
Lakartidningen
[Self-administered remifentanil during labor. Satisfied mothers despite moderate pain relief, as shown in quality assurance project].
PMID 24437099
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R1908
Remifentanil hydrochloride, ≥97% (HPLC), powder
C20H28N2O5·HCl