EMAIL THIS PAGE TO A FRIEND
Klinicheskaia meditsina
[Anti-inflammatory potential of melatonin].
PMID 24437164
Abstract
The summary of anti-inflammatory properties, cellular and systemic mechanisms of action of epiphyseal melatonin is presented. Dual effect of interaction between melatonin and non-steroidal anti-inflammatory agents is discussed with reference to enhancement of their specific activity and prevention of side effects.