EMAIL THIS PAGE TO A FRIEND
Annals of hematology
Absolute lymphocyte count and risk of short-term infection in patients with immune thrombocytopenia.
PMID 24441917
Abstract
Patients with immune thrombocytopenia (ITP) may be at increased risk of infection because of the steroids and other immunosuppressive agents used in its treatment. This study aimed to identify events that are associated with infection within 6xa0months of diagnosis and the impact that infection has on survival. We retrospectively evaluated 239 patients (107 men, 132 women; median age 61xa0years) diagnosed between January 1997 and August 2011. Every patient received steroid treatment according to the platelet count and the extent of bleeding. Logistic regression analysis was used to identify risk factors associated with the development of infection within 6xa0months of ITP being diagnosed. Sixty-two patients (25.9xa0%) developed an infection within 6xa0months of diagnosis. Multivariate analysis revealed that a lower absolute lymphocyte count (ALC) at diagnosis (<1 × 10(9)/l) was an independent risk factor for infection (P = 0.039; 95xa0% confidence interval, 1.033-3.599; odds ratio, 1.928). The time to infection event is significant shorter in those of low ALC, compared with those of higher ALC (P = 0.032). Furthermore, the 1-year mortality rate after ITP diagnosis was significantly higher in those patients who developed an infection (P = 0.001). ITP patients with a low absolute lymphocyte count at diagnosis have an increased risk of infection, and those who develop infections have lower 1-year survival.