EMAIL THIS PAGE TO A FRIEND
FEBS letters
The chaperone FdsC for Rhodobacter capsulatus formate dehydrogenase binds the bis-molybdopterin guanine dinucleotide cofactor.
PMID 24444607
Abstract
Molybdoenzymes are complex enzymes in which the molybdenum cofactor (Moco) is deeply buried in the enzyme. Most molybdoenzymes contain a specific chaperone for the insertion of Moco. For the formate dehydrogenase FdsGBA from Rhodobacter capsulatus the two chaperones FdsC and FdsD were identified to be essential for enzyme activity, but are not a subunit of the mature enzyme. Here, we purified and characterized the FdsC protein after heterologous expression in Escherichia coli. We were able to copurify FdsC with the bound Moco derivate bis-molybdopterin guanine dinucleotide. This cofactor successfully was used as a source to reconstitute the activity of molybdoenzymes.