EMAIL THIS PAGE TO A FRIEND
Antonie van Leeuwenhoek
Indole-3-acetic acid in plant-microbe interactions.
PMID 24445491
Abstract
Indole-3-acetic acid (IAA) is an important phytohormone with the capacity to control plant development in both beneficial and deleterious ways. The ability to synthesize IAA is an attribute that many bacteria including both plant growth-promoters and phytopathogens possess. There are three main pathways through which IAA is synthesized; the indole-3-pyruvic acid, indole-3-acetamide and indole-3-acetonitrile pathways. This chapter reviews the factors that effect the production of this phytohormone, the role of IAA in bacterial physiology and in plant-microbe interactions including phytostimulation and phytopathogenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
286281
Indole-3-acetamide, 98%
C10H10N2O