EMAIL THIS PAGE TO A FRIEND
Releve epidemiologique hebdomadaire
Schistosomiasis: number of people receiving preventive chemotherapy in 2012.
PMID 24446558
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P4668
Praziquantel, anthelminic, neurogenic
C19H24N2O2
46648
Praziquantel, VETRANAL, analytical standard
C19H24N2O2