EMAIL THIS PAGE TO A FRIEND
Mucosal immunology
IL-17-producing NKT cells depend exclusively on IL-7 for homeostasis and survival.
PMID 24448098
Abstract
Natural killer T (NKT) cells are innate-like T cells that rapidly recognize pathogens and produce cytokines that shape the ensuing immune response. IL-17-producing NKT cells are enriched in barrier tissues, such as the lung, skin, and peripheral lymph nodes, and the factors that maintain this population in the periphery have not been elucidated. Here we show that NKT17 cells deviate from other NKT cells in their survival requirements. In contrast to conventional NKT cells that are maintained by IL-15, RORγt(+) NKT cells are IL-15 independent and instead rely completely on IL-7. IL-7 initiates a T-cell receptor-independent (TCR-independent) expansion of NKT17 cells, thus supporting their homeostasis. Without IL-7, survival is dramatically impaired, yet residual cells remain lineage committed with no downregulation of RORγt evident. Their preferential response to IL-7 does not reflect enhanced signaling through STAT proteins, but instead is modulated via the PI3K/AKT/mTOR signaling pathway. The ability to compete for IL-7 is dependent on high-density IL-7 receptor expression, which would promote uptake of low levels of IL-7 produced in the non-lymphoid sites of lung and skin. This dependence on IL-7 is also reported for RORγt(+) innate lymphoid cells and CD4(+) Th17 cells, and suggests common survival requirements for functionally similar cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1400420
Anti-CD160 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB1401560
Anti-CD160 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2105461
Anti-CD160 antibody produced in rabbit, affinity isolated antibody
SAB4500215
Anti-CD160 antibody produced in rabbit, affinity isolated antibody
SAB4700041 Anti-CD3, low endotoxin antibody, Mouse monoclonal, clone OKT3, purified immunoglobulin, buffered aqueous solution
SAB4300686
Anti-CD3E antibody produced in rabbit, affinity isolated antibody
H7157 Anti-Hepatocyte Growth Factor antibody produced in goat, affinity isolated antibody, lyophilized powder
H0652 Anti-Hepatocyte Growth Factor antibody produced in goat, IgG fraction of antiserum, lyophilized powder
H0540 Anti-Hepatocyte Growth Factor antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB2101033
Anti-HGF (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2101034
Anti-HGF (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2105571
Anti-HGF antibody produced in rabbit, affinity isolated antibody
HPA040360
Anti-HGF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA044088
Anti-HGF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410624
Anti-HGF antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1406025
Anti-IL7 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA019590
Anti-IL7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410709
Anti-IL7 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
GW22461F Anti-Interleukin-2 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
I8398 Anti-Interleukin-2 antibody produced in goat, IgG fraction of antiserum, lyophilized powder
I8273 Anti-Interleukin-2 antibody produced in goat, IgG fraction of antiserum, lyophilized powder
I7401 Anti-Interleukin-2 antibody produced in goat, affinity isolated antibody, lyophilized powder
I7784 Anti-Interleukin-2 antibody produced in goat, affinity isolated antibody, lyophilized powder
S1542 Anti-Substance P antibody produced in rabbit, whole antiserum
HPA014429 Anti-TAC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1408632
Anti-TAC1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
C7048
Monoclonal Anti-CD3 antibody produced in mouse, clone UCHT-1, purified immunoglobulin, buffered aqueous solution
SAB4700040 Monoclonal Anti-CD3 antibody produced in mouse, clone OKT3, purified immunoglobulin, buffered aqueous solution
SAB4700044 Monoclonal Anti-CD3 antibody produced in mouse, clone MEM-57, purified immunoglobulin, buffered aqueous solution
SAB4700045 Monoclonal Anti-CD3 antibody produced in mouse, clone MEM-92, purified immunoglobulin, buffered aqueous solution