EMAIL THIS PAGE TO A FRIEND
Environmental pollution (Barking, Essex : 1987)
Occurrence and spatial distribution of microplastics in sediments from Norderney.
PMID 24448461
Abstract
The spatial distribution of small potential microplastics (SPM) (<1 mm) in beach sediments was studied on a 500 m stretch of the North Sea island of Norderney. Their correlation with visible plastic debris (VPD) (>1 mm) was also examined. Small microparticles were extracted from 36 one kg sediment samples and analysed by visual microscopic inspection and partly by thermal desorption pyrolysis gas chromatography/mass spectrometry. The smallest particle size that could be analysed with this method was estimated to be 100 μm. The mean number of SPM at the three sampling sites (n = 12) was 1.7, 1.3 and 2.3 particles per kg dry sediment, respectively. SPM were identified as polypropylene, polyethylene, polyethylene terephthalate, polyvinylchloride, polystyrene and polyamide. The organic plastic additives found were benzophenone, 1,2-benzenedicarboxylic acid, dimethyl phthalate, diethylhexyl phthalate, dibutyl phthalate, diethyl phthalate, phenol and 2,4-di-tert-butylphenol. Particles were distributed rather homogenously and the occurrence of SPM did not correlate with that of VPD.