EMAIL THIS PAGE TO A FRIEND
Environmental pollution (Barking, Essex : 1987)
Components from wheat roots modify the bioactivity of ZnO and CuO nanoparticles in a soil bacterium.
PMID 24448483
Abstract
ZnO and CuO nanoparticles (NPs) have widespread commercial uses and their impact on agricultural systems is unresolved. This study examined whether the metabolites washed from wheat (Triticum aestivum L.) roots modulated the metabolic response to the NPs of a biosensor generated in the root colonizer, Pseudomonas putida KT2440. The root wash components boosted light output of the biosensor consistent with their catabolism. Dose-dependent and rapid inhibition of cell metabolism occurred with both ZnO and CuO NPs in water suspensions but high light output was maintained in root wash. Root wash also protected biosensor output in challenges with Zn ions. However the root wash components did not protect culturability or biosensor light output upon exposure to Cu ions. Imaging by atomic force microscopy suggested that root wash materials coated the NPs. We deduced that the response of a microbe to these metal oxide NPs could be negated by components released from roots.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
203130
Copper(II) oxide, 99.999% trace metals basis
CuO
450804
Copper(II) oxide, powder, 99.995% trace metals basis
CuO
450812
Copper(II) oxide, powder, 99.99% trace metals basis
CuO
208841
Copper(II) oxide, powder, <10 μm, 98%
CuO
544868
Copper(II) oxide, nanopowder, <50 nm particle size (TEM)
CuO
241741
Copper(II) oxide, ACS reagent, ≥99.0%
CuO
310433
Copper(II) oxide, needles, mixture of CuO and Cu2O, ACS reagent
CuO
61200
Copper(II) oxide, puriss. p.a., ≥99.0% (RT), powder
CuO
94737
Copper(II) oxide, ≥99.0% (RT), granular
CuO
417971
Copper(II) oxide on alumina, 14-20 mesh, extent of labeling: 13 wt. % loading
CuO
96479
Zinc oxide, puriss. p.a., ACS reagent, ≥99.0% (KT)
OZn
14439
Zinc oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. for dried substance)
OZn
96492
Zinc oxide, tested according to Ph.Eur.
OZn
93632
Zinc oxide, analytical standard
OZn
204951
Zinc oxide, 99.999% trace metals basis
OZn
255750
Zinc oxide, 99.99% trace metals basis
OZn
205532
Zinc oxide, ReagentPlus®, powder, <5 μm particle size, 99.9%
OZn
544906
Zinc oxide, nanopowder, <100 nm particle size
OZn
677450
Zinc oxide, nanopowder, <50 nm particle size (BET), >97%
OZn
752681
Zinc oxide, sputtering target, diam. × thickness 3.00 in. × 0.125 in., 99.99% trace metals basis
OZn
773999
Zinc oxide, nanowires, diam. × L 90 nm × 1 μm
OZn
773980
Zinc oxide, nanowires, diam. × L 50 nm × 300 nm
OZn
774006
Zinc oxide, nanowires, diam. × L 300 nm × 4-5 μm
OZn
721077
Zinc oxide, dispersion, nanoparticles, <100 nm particle size (TEM), ≤40 nm avg. part. size (APS), 20 wt. % in H2O
721085
Zinc oxide, dispersion, nanoparticles, 40 wt. % in ethanol, <130 nm particle size
721093
Zinc oxide, dispersion, nanoparticles, <110 nm particle size, 40 wt. % in butyl acetate