EMAIL THIS PAGE TO A FRIEND
Arthritis & rheumatology (Hoboken, N.J.)
An essential role of caspase 1 in the induction of murine lupus and its associated vascular damage.
PMID 24449582
Abstract
Systemic lupus erythematosus (SLE) is a systemic autoimmune syndrome associated with organ damage and an elevated risk of cardiovascular disease resulting from activation of both innate and adaptive immune pathways. Recently, increased activation of the inflammasome machinery in SLE has been described. Using the mouse model of pristane-induced lupus, we undertook this study to explore whether caspase 1, the central enzyme of the inflammasome, plays a role in the development of SLE and its associated vascular dysfunction. Eight-week-old wild-type (WT) or caspase 1(-/-) mice were injected intraperitoneally with phosphate buffered saline or pristane. Six months after injection, mice were euthanized, and the development of a lupus phenotype and vascular dysfunction was assessed. While WT mice exposed to pristane developed autoantibodies and a strong type I interferon response, mice lacking caspase 1 were significantly protected against these features as well as against pristane-induced vascular dysfunction. Further, the development of immune complex glomerulonephritis, which was prominent after pristane exposure in WT mice, was significantly abrogated in caspase 1(-/-) mice. These results indicate that caspase 1 is an essential component in the development of lupus and its associated vascular dysfunction and that it may play an important role in the cross-talk between environmental exposures and autoimmunity development, thus identifying a novel pathway for therapeutic targeting.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2501632
Anti-BST2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1400028
Anti-BST2 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB4500221
Anti-BST2 antibody produced in rabbit, affinity isolated antibody
HPA017060
Anti-BST2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300654
Anti-CD11c antibody produced in rabbit, affinity isolated antibody
GW23003 Anti-CD19 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA004252
Anti-CD4 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
RAB0267
Human IL-18 BPa  ELISA Kit, for serum, plasma, cell culture supernatant and urine
RAB0268
Human IL-18 R β ELISA Kit, for serum, plasma, cell culture supernatant and urine
RAB0543
Human IL18 / Interleukin-18 ELISA Kit, for serum, plasma, cell culture supernatants and urine
SAB1403618
Monoclonal Anti-BST2 antibody produced in mouse, clone 2E6, purified immunoglobulin, buffered aqueous solution
SAB4700390 Monoclonal Anti-CD11c antibody produced in mouse, clone BU15, purified immunoglobulin, buffered aqueous solution
SAB4700108 Monoclonal Anti-CD19 antibody produced in mouse, clone 4G7, purified immunoglobulin, buffered aqueous solution
SAB4700109 Monoclonal Anti-CD19 antibody produced in mouse, clone LT19, purified immunoglobulin, buffered aqueous solution
SAB1403645
Monoclonal Anti-CD19 antibody produced in mouse, clone 1C9, purified immunoglobulin, buffered aqueous solution
SAB4700059
Monoclonal Anti-CD4 , (N-terminal) antibody produced in mouse, clone MEM-241, purified immunoglobulin, buffered aqueous solution
C1805 Monoclonal Anti-CD4 antibody produced in mouse, clone Q4120, purified from hybridoma cell culture
SAB4100489
Monoclonal Anti-CD4 antibody produced in mouse, clone CD4 -A409, culture supernatant
SAB4700054 Monoclonal Anti-CD4 antibody produced in mouse, clone MEM-16, purified immunoglobulin, buffered aqueous solution
SAB4700065
Monoclonal Anti-CD4 antibody produced in mouse, clone MEM-115, purified immunoglobulin, buffered aqueous solution
SAB4700733 Monoclonal Anti-CD4 antibody produced in mouse, clone OX-35, purified immunoglobulin, buffered aqueous solution
AMAB90754
Monoclonal Anti-CD4 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0395, purified immunoglobulin, buffered aqueous glycerol solution
RAB0810 Mouse Il18 / Interleukin-18 ELISA Kit, for serum, plasma and cell culture supernatants
P2870
Pristane, synthetic, ≥98% (GC)
C19H40
P9622
Pristane, synthetic, liquid, sterile-filtered, BioReagent
C19H40
WH0000930M1
Monoclonal Anti-CD19 antibody produced in mouse, clone 1G3, purified immunoglobulin, buffered aqueous solution
PRS4661
Anti-BST2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1400029
Anti-BST2 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution