EMAIL THIS PAGE TO A FRIEND
Journal of integrative plant biology
Overexpression of OsKTN80a, a katanin P80 ortholog, caused the repressed cell elongation and stalled cell division mediated by microtubule apparatus defects in primary root in Oryza sativa.
PMID 24450597