EMAIL THIS PAGE TO A FRIEND
ACS applied materials & interfaces
Clean method for the synthesis of reduced graphene oxide-supported PtPd alloys with high electrocatalytic activity for ethanol oxidation in alkaline medium.
PMID 24451011
Abstract
In this article, a clean method for the synthesis of PtPd/reduced graphene oxide (RGO) catalysts with different Pt/Pd ratios is reported in which no additional components such as external energy (e.g., high temperature or high pressure), surfactants, or stabilizing agents are required. The obtained catalysts were characterized by X-ray diffraction (XRD), transmission electron microscopy (TEM), high-resolution transmission electron microscopy (HRTEM), Raman spectroscopy, X-ray photoelectron spectroscopy (XPS), induced coupled plasma atomic emission spectroscopy (ICP-AES), and electrochemical measurements. The HRTEM measurements showed that all of the metallic nanoparticles (NPs) exhibited well-defined crystalline structures. The composition of these Pt-Pd/RGO catalysts can be easily controlled by adjusting the molar ratio of the Pt and Pd precursors. Both cyclic voltammetry (CV) and chronoamperometry (CA) results demonstrate that bimetallic PtPd catalysts have superior catalytic activity for the ethanol oxidation reaction compared to the monometallic Pt or Pd catalyst, with the best performance found with the PtPd (1:3)/RGO catalyst. The present study may open a new approach for the synthesis of PtPd alloy catalysts, which is expected to have promising applications in fuel cells.
Related Materials