EMAIL THIS PAGE TO A FRIEND
Biomedical chromatography : BMC
A rapid, simple and reliable HPLC-triple quadrupole tandem mass spectrometer method for a simultaneous quantification of irinotecan and its active metabolite 7-ethyl-10-hydroxycamptothecin (SN38) in mouse plasma.
PMID 24458571
Abstract
A simple, fast and reliable high-performance liquid chromatography-triple quadrupole mass spectrometry method (HPLC-MS/MS method) was developed, validated and used for the simultaneous quantification of irinotecan and 7-ethyl-10-hydroxycamptothecin (SN38) in heparinized mouse plasma. Camptothecin was used as the internal standard. A single-step protein precipitation without evaporation and reconstitution steps was adopted as sample processing method. Our bioanalytical method was validated in compliance with the guidelines from the European Medicines Agency. The lower limit of quantification for both irinotecan and SN38 was 5 ng/mL. The calibration curves for both analytes fitted to a 1/x(2) weighted linear regression model and ranged from 5 to 1000 ng/mL. The intra-run and inter-run precisions were within 8.6%, and the intra-run and inter-run accuracies were within 96.4-103.9%. Our validated bioanalytical method was successfully applied to the pharmacokinetic study in mice, in which 4 mg/kg irinotecan was intraperitoneally injected.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I1406
Irinotecan hydrochloride, topoisomerase inhibitor
C33H38N4O6 · HCl