EMAIL THIS PAGE TO A FRIEND
Environmental toxicology
Ameliorative effect of polyphenols from Padina boergesenii against ferric nitrilotriacetate induced renal oxidative damage: With inhibition of oxidative hemolysis and in vitro free radicals.
PMID 24458998
Abstract
The aim of this study was to evaluate the antioxidant activities of diethyl ether (DEE) and methanol (M) extracts from brown alga Padina boergesenii using in vitro and in vivo antioxidant assay, which may help to relate the antioxidant properties with the possible outline of its ameliorative effect. M extract showed higher radical scavenging activity through ferric reducing antioxidant power 139.11 µmol tannic acid equivalent/g; DPPH 71.32 ± 0.56%; deoxyribose radical 88.31 ± 0.47%, and total antioxidant activity 0.47 ± 0.02 mg ascorbic acid equivalents/g. Oxidative red blood cell (RBC) hemolysis inhibition rate was significantly higher in M extract (150 mg/kg body weight) in reference to total phenolic content (r = 0.935). Rats administered with DEE and M extracts (150 mg/kg body weight) for seven days before the administration of ferric nitrilotriacetate (9 mg of Fe/mg/kg bodyweight). Rats pretreated with extracts significantly changed the level of renal microsomal lipid peroxidation, glutathione, and antioxidant enzymes in post-mitochondrial supernatant (P < 0.05). Ameliorative effect of extracts against renal oxidative damage was evident in rat kidney through changes in necrotic and epithelial cells. HPTLC technique has identified the presence of rutin with reference to retardation factor (Rf ) in both the extracts. These findings support the source of polyphenols (rutin) from P. boergesenii had potent antioxidant activity; further work on isolation of bioactive compounds can be channeled to develop as a natural antioxidant.