EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutical compounding
Methocarbamol suspension for the treatment of rhabdomyolysis in equines.
PMID 24459784
Abstract
Rhabdomyolysis in equines occurs in horses due to physical overexertion or underlying pathologic myopathy. Methocarbamol is a muscle relaxant that can be used in equines to treat symptoms associated with Rhabdomyolysis. Methocarbamol is available as a solution for injection but is not commercially available as an oral suspension. This article focuses on the treatment of Tying-up caused by overexertion, and details the treatment of Rhabdomyolysis with an oral suspension that was prepared for a veterinarian by a compounding pharmacist.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G7012
Guaiacol glyceryl ether carbamate, analytical standard
C11H15NO5