EMAIL THIS PAGE TO A FRIEND
Journal of Cellular and Molecular Medicine
Engineering of pre-vascularized urethral patch with muscle flaps and hypoxia-activated hUCMSCs improves its therapeutic outcome.
PMID 24460735