EMAIL THIS PAGE TO A FRIEND
The American journal of emergency medicine
Pulmonary thromboembolism due to paliperidone: report of 2 cases.
PMID 24462396
Abstract
Venous thromboembolism (VTE) is serious medical condition, which might be caused by psychotropic medications. Previously, antipsychotic-induced VTE due to olanzapine, risperidone, clozapine, and amisulpiride was reported. In this report, we present 2 cases of paliperidone-induced VTE.