EMAIL THIS PAGE TO A FRIEND
Testing
Dosage and cell line dependent inhibitory effect of bFGF supplement in human pluripotent stem cell culture on inactivated human mesenchymal stem cells.
PMID 24465853