EMAIL THIS PAGE TO A FRIEND
Current cancer drug targets
Cetuximab inhibits gastric cancer growth in vivo, independent of KRAS status.
PMID 24467518
Abstract
Mutation status of Kirsten rat sarcoma viral oncogene homolog (KRAS) may serve as a negative predictive marker of Cetuximab in treating colorectal cancer. The present study was to determine the role of KRAS status in EGFR antibody treatment for gastric cancer. KRAS status was clarified in SGC-7901 (wild type) and YCC-2 (G→A mutation) gastric cancer cell lines. Anti-proliferative and pro-apoptotic effects of Cetuximab were tested both in vitro and in vivo, the expression of phosphorylated (p) ERK, a downstream protein in EGFR-RAS-MEK pathway, was also analyzed. No significant in vitro anti-proliferative or pro-apoptotic effects of Cetuximab were observed in both cell lines. The growth of either SGC-7901 or YCC-2 gastric cancer xenograft was significantly inhibited by Cetuximab. Apoptosis was induced in SGC-7901 but not in YCC-2 xenografts after Cetuximab treatment. The expression of pERK was up-regulated in YCC-2 but not SGC-7901 xenografts after Cetuximab treatment. In conclusion, KRAS (G→A) mutation does not affect in vivo anti-cancer efficacy of Cetuximab.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300352
Anti-EGFR (Ab-1070) antibody produced in rabbit, affinity isolated antibody
SAB4300353
Anti-EGFR (Ab-1092) antibody produced in rabbit, affinity isolated antibody
SAB4300510
Anti-EGFR (Ab-1110) antibody produced in rabbit, affinity isolated antibody
SAB4300472
Anti-EGFR (Ab-1172) antibody produced in rabbit, affinity isolated antibody
SAB4300602
Anti-EGFR (Ab-1197) antibody produced in goat, affinity isolated antibody
SAB4300454
Anti-EGFR (Ab-693) antibody produced in rabbit, affinity isolated antibody
GW21146 Anti-EGFR antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1405746
Anti-EGFR antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA018530
Anti-EGFR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA001200
Anti-EGFR antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4500711
Anti-EGFR antibody produced in rabbit, affinity isolated antibody
SAB4500712
Anti-EGFR antibody produced in rabbit, affinity isolated antibody
E1282 Anti-Epidermal Growth Factor Receptor antibody produced in goat, affinity isolated antibody, lyophilized powder
WH0001956M2
Monoclonal Anti-EGFR antibody produced in mouse, clone 4H2, purified immunoglobulin, buffered aqueous solution
AMAB90816
Monoclonal Anti-EGFR antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0815, purified immunoglobulin, buffered aqueous glycerol solution
E2760
Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse, clone 29.1, ascites fluid
E3138
Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse, clone F4, ascites fluid
E0532 Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse, clone 102618, purified immunoglobulin, lyophilized powder
PLA0101
Rabbit anti-EGFR Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
SAB4300453
Anti-EGFR (Ab-678) antibody produced in rabbit, affinity isolated antibody
SAB4300480
Anti-EGFR (Ab-1197) antibody produced in rabbit, affinity isolated antibody
SAB4300481
Anti-EGFR (Ab-869) antibody produced in rabbit, affinity isolated antibody
SAB4500720
Anti-EGFR antibody produced in rabbit, affinity isolated antibody