EMAIL THIS PAGE TO A FRIEND
Cancer letters
Mevalonate metabolism in cancer.
PMID 24467965
Abstract
Cancer cells are characterized by sustained proliferative signaling, insensitivity to growth suppressors and resistance to apoptosis as well as by replicative immortality, the capacity to induce angiogenesis and to perform invasive growth. Additional hallmarks of cancer cells include the reprogramming of energy metabolism as well as the ability to evade immune surveillance. The current review focuses on the metabolic reprogramming of cancer cells and on the immune system's capacity to detect such changes in cancer cell metabolism. Specifically, we focus on mevalonate metabolism, which is a target for drug and immune based cancer treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
90469
(RS)-Mevalonic acid lithium salt, ≥96.0% (GC)
C6H11O4 · xLi+
42147
(RS)-Mevalonic acid lithium salt, analytical standard
C6H11O4 · xLi+