EMAIL THIS PAGE TO A FRIEND
Metal ions in life sciences
Nickel and human health.
PMID 24470096
Abstract
This review focuses on the impact of nickel on human health. In particular, the dual nature of nickel as an essential as well as toxic element in nature is described, and the main forms of nickel that can come in contact with living systems from natural sources and anthropogenic activities are discussed. Concomitantly, the main routes of nickel uptake and transport in humans are covered, and the potential dangers that nickel exposure can represent for health are described. In particular, the insurgence of nickel-derived allergies, nickel-induced carcinogenesis as well as infectious diseases caused by human pathogens that rely on nickel-based enzymes to colonize the host are reviewed at different levels, from their macroscopic aspects on human health to the molecular mechanisms underlying these points. Finally, the importance of nickel as a beneficial element for human health, especially being essential for microorganisms that colonize the human guts, is examined.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
357553
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Ni
267007
Nickel, foil, thickness 0.25 mm, 99.995% trace metals basis
Ni
268259
Nickel, foil, thickness 0.125 mm, ≥99.9%
Ni
357588
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Ni
203904
Nickel, powder, <150 μm, 99.99% trace metals basis
Ni
266981
Nickel, powder, <50 μm, 99.7% trace metals basis
Ni
268283
Nickel, powder, <1 μm, 99.8% trace metals basis
Ni
577995
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Ni
267074
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Ni
267058
Nickel, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Ni
357626
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Ni
357634
Nickel, wire, diam. 0.25 mm, ≥99.9%
Ni
767484
Nickel, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis
Ni
GF82156912
Nickel, rod, 100mm, diameter 12.0mm, 99%
Ni
GF11598194
Nickel, rod, 100mm, diameter 12.7mm, 99.98%
Ni
GF29576010
Nickel, rod, 100mm, diameter 16mm, 99.99+%
Ni
GF83584143
Nickel, rod, 100mm, diameter 2.0mm, 99.99+%
Ni
GF45193697
Nickel, rod, 100mm, diameter 2.0mm, annealed, 99%
Ni
GF11517948
Nickel, rod, 100mm, diameter 20.0mm, 99%
Ni
GF39205931
Nickel, rod, 100mm, diameter 25.4mm, 99.98%
Ni
GF69167920
Nickel, rod, 100mm, diameter 3.0mm, 99%
Ni
GF70332870
Nickel, rod, 100mm, diameter 3.3mm, 99.99+%
Ni
GF99799968
Nickel, rod, 100mm, diameter 30mm, 99%
Ni
GF90754241
Nickel, rod, 100mm, diameter 40mm, 99%
Ni
GF20561385
Nickel, rod, 100mm, diameter 5.0mm, 99.99+%
Ni
GF90932823
Nickel, rod, 100mm, diameter 6.35mm, 99%
Ni
GF50593002
Nickel, rod, 100mm, diameter 9.5mm, 99%
Ni
GF68355085
Nickel, rod, 200mm, diameter 1.5mm, 99.99+%
Ni
GF66250256
Nickel, rod, 200mm, diameter 10.0mm, 99.99+%
Ni
GF69613825
Nickel, rod, 200mm, diameter 12.0mm, 99%
Ni