EMAIL THIS PAGE TO A FRIEND
Journal of Chromatographic Science
Development of a new method for the identification of degradation products of V-type nerve agents by liquid chromatography-tandem mass spectrometry.
PMID 24470166