EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Human hepatic lipase. Cloned cDNA sequence, restriction fragment length polymorphisms, chromosomal localization, and evolutionary relationships with lipoprotein lipase and pancreatic lipase.
PMID 2447084
Abstract
Human hepatic lipase is an important enzyme in high density lipoprotein (HDL) metabolism, being implicated in the conversion of HDL2 to HDL3. Three human hepatic lipase cDNA clones were identified in two lambda gt11 libraries from human liver. The cDNA-derived amino acid sequence predicts a protein of 476 amino acid residues, preceded by a 23-residue signal peptide. Four potential N-glycosylation sites are identified, two of which are conserved in rat hepatic lipase. On alignment with human, mouse, and bovine lipoprotein lipase, the same two sites were also conserved in lipoprotein lipase in all three species. Stringent conservation of the cysteine residues was also evident. Comparative analysis of amino acid sequences shows that hepatic lipase evolves at a rapid rate, 2.07 x 10(-9) substitutions/site/year, about four times that in lipoprotein lipase and half that in pancreatic lipase. Further, hepatic lipase and pancreatic lipase appear to be evolutionarily closer to each other than either of them is to lipoprotein lipase. Southern blot analysis revealed high frequency restriction fragment length polymorphisms of the hepatic lipase gene for the enzymes HindIII and MspI. these polymorphisms will be useful for haplotype and linkage analysis of the hepatic lipase gene. Using cloned human hepatic lipase cDNA as a hybridization probe, we performed Southern blot analysis of a panel of 13 human-rodent somatic cell hybrids. Concordance analysis of the various hybrid clones indicates that the hepatic lipase gene is located on the long arm of human chromosome 15. Analysis of hybrids containing different translocations of chromosome 15 localized the gene to the region 15q15----q22.