EMAIL THIS PAGE TO A FRIEND
The American journal of medicine
An absolute obstacle: cardiac metastasis of synovial sarcoma.
PMID 24472446
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I0060000
Ifosfamide, European Pharmacopoeia (EP) Reference Standard
C7H15Cl2N2O2P