EMAIL THIS PAGE TO A FRIEND
Revista da Sociedade Brasileira de Medicina Tropical
Dengue infection in children and adolescents: clinical profile in a reference hospital in northeast Brazil.
PMID 24474020
Abstract
This study aimed to describe the clinical spectrum of dengue in children and adolescents from a hyperendemic region who were admitted for hospitalization. A retrospective study was conducted on patients diagnosed with dengue infection upon admission to a reference center in Fortaleza, Brazil. Of the 84 patients included, 42 underwent confirmatory testing. The main symptoms were fever, abdominal pain and vomiting. The median level of serum aspartate aminotransferase was 143.5±128mg/dL. A peculiar clinical profile was evident among children and adolescents with dengue infection in a reference center in northeast Brazil, including gastrointestinal symptoms and liver involvement.