EMAIL THIS PAGE TO A FRIEND
Chembiochem : a European journal of chemical biology
Active efflux influences the potency of quorum sensing inhibitors in Pseudomonas aeruginosa.
PMID 24478193
Abstract
Many bacteria regulate gene expression through a cell-cell signaling process called quorum sensing (QS). In proteobacteria, QS is largely mediated by signaling molecules known as N-acylated L-homoserine lactones (AHLs) and their associated intracellular LuxR-type receptors. The design of non-native small molecules capable of inhibiting LuxR-type receptors (and thereby QS) in proteobacteria is an active area of research, and numerous lead compounds are AHL derivatives that mimic native AHL molecules. Much of this previous work has focused on the pathogen Pseudomonas aeruginosa, which controls an arsenal of virulence factors and biofilm formation through QS. The MexAB-OprM efflux pump has been shown to play a role in the secretion of the major AHL signal in P. aeruginosa, N-(3-oxododecanoyl) L-homoserine lactone. In the current study, we show that a variety of non-native AHLs and related derivatives capable of inhibiting LuxR-type receptors in P. aeruginosa display significantly higher potency in a P. aeruginosa Δ(mexAB-oprM) mutant, suggesting that MexAB-OprM also recognizes these compounds as substrates. We also demonstrate that the potency of 5,6-dimethyl-2-aminobenzimidazole, recently shown to be a QS and biofilm inhibitor in P. aeruginosa, is not affected by the presence/absence of the MexAB-OprM pump. These results have implications for the use of non-native AHLs and related derivatives as QS modulators in P. aeruginosa and other bacteria, and provide a potential design strategy for the development of new QS modulators that are resistant to active efflux.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H6515
L-Homoserine
C4H9NO3