EMAIL THIS PAGE TO A FRIEND
Journal of Veterinary Pharmacology and Therapeutics
Pharmacokinetics of doxycycline in tilapia (Oreochromis aureus ? Oreochromis niloticus) after intravenous and oral administration.
PMID 24479741