EMAIL THIS PAGE TO A FRIEND
The Journal of Biological Chemistry
Phosphorylation of myosin II-interacting guanine nucleotide exchange factor (MyoGEF) at threonine 544 by aurora B kinase promotes the binding of polo-like kinase 1 to MyoGEF.
PMID 24482237