EMAIL THIS PAGE TO A FRIEND
Journal of oral pathology & medicine : official publication of the International Association of Oral Pathologists and the American Academy of Oral Pathology
Toll-like receptor 9 expression in mucoepidermoid salivary gland carcinoma may associate with good prognosis.
PMID 24484266
Abstract
Mucoepidermoid carcinoma (MEC) is the most common salivary gland malignancy. Although several biomarkers have been evaluated, histological grade remains the most valuable prognostic marker. Toll-like receptor 9 (TLR9) is an immune receptor recognizing microbial DNA. Its expression associates with prognosis or cancer properties in several cancers. This study examined the role of TLR9 in MEC. Sixty patients with salivary gland MEC were collected from two Finnish university hospitals, and tumor samples were stained for TLR9. Salivary gland high-grade MEC cell line (UT-MUC-1) was cultured to assess TLR9 and MMP-13 expression. The function of TLR9 was studied in vitro using traditional Matrigel(®) invasion assay and novel human myoma organotypic model. Cancer-specific survival was related with tumor grade (P = 0.01), and there were no deaths in patients with low-grade MEC. TLR9 was expressed in 56 of 60 (93%) tumors. High TLR9 expression indicated better survival in the patient series (P = 0.002) and showed a trend for association with lower disease stage (P = 0.06) and higher differentiation grade (P = 0.068). In multivariate analysis, TLR9 expression was prognostically insignificant due to heavy correlation to disease stage and higher gradus. Treating UT-MUC-1 cells with TLR9 ligand CpG in vitro induced MMP-13 expression and invasion in Matrigel(®) invasion assay, whereas decreased invasion was seen in myoma organotypic model. Functional TLR9 is present in salivary MEC, and high level of expression may indicate good prognosis. However, more studies are needed to evaluate biological consequences of TLR9 interaction in tumor cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA000451
Anti-MKI67 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA001164
Anti-MKI67 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
GW22790 Anti-TLR9 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1412111 ANTI-TLR9 antibody produced in mouse, clone 2D11, purified immunoglobulin, buffered aqueous solution
SAB1412155
ANTI-TLR9 antibody produced in mouse, clone 3B7, purified immunoglobulin, buffered aqueous solution
SAB1412156
ANTI-TLR9 antibody produced in mouse, clone 2H5, purified immunoglobulin, buffered aqueous solution
SAB1412299
ANTI-TLR9 antibody produced in mouse, clone 1E4, purified immunoglobulin, buffered aqueous solution
SAB1412300
ANTI-TLR9 antibody produced in mouse, clone 2G3, purified immunoglobulin, buffered aqueous solution
SAB1412301
ANTI-TLR9 antibody produced in mouse, clone 1D3, purified immunoglobulin, buffered aqueous solution
SAB1412302
ANTI-TLR9 antibody produced in mouse, clone 2C3, purified immunoglobulin, buffered aqueous solution
SAB3500313
Anti-TLR9 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2104136
Anti-TLR9, (N-terminal) antibody produced in rabbit, affinity isolated antibody
WH0004288M1
Monoclonal Anti-MKI67 antibody produced in mouse, clone 7B8, purified immunoglobulin, buffered aqueous solution
AMAB90870
Monoclonal Anti-MKI67 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1234, purified immunoglobulin, buffered aqueous glycerol solution
SAB5300423 Monoclonal Anti-MKI67 antibody produced in mouse, clone 4A1, ascites fluid
SAB5300424 Monoclonal Anti-MKI67 antibody produced in mouse, clone 9C12B2, ascites fluid
SAB5300425 Monoclonal Anti-MKI67 antibody produced in mouse, clone 8D5, ascites fluid
WH0054106M3
Monoclonal Anti-TLR9 antibody produced in mouse, clone 1E8, purified immunoglobulin, buffered aqueous solution
SAB1405118
Monoclonal Anti-TLR9 antibody produced in mouse, clone 1F4, purified immunoglobulin, buffered aqueous solution