EMAIL THIS PAGE TO A FRIEND
Molecular breeding : new strategies in plant improvement
The control of seed oil polyunsaturate content in the polyploid crop species
PMID 24489479
Abstract
Many important plant species have polyploidy in their recent ancestry, complicating inferences about the genetic basis of trait variation. Although the principal locus controlling the proportion of polyunsaturated fatty acids (PUFAs) in seeds of
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
RAP-RO rAPid Alkaline Phosphatase