EMAIL THIS PAGE TO A FRIEND
Journal of clinical oncology : official journal of the American Society of Clinical Oncology
BRAF V600E mutation in two distinct meningeal melanocytomas associated with a nevus of Ota.
PMID 24493731