EMAIL THIS PAGE TO A FRIEND
Journal of environmental sciences (China)
Diel and seasonal variation of methane and carbon dioxide fluxes at site Guojiaba, the Three Gorges Reservoir.
PMID 24494493
Abstract
In order to investigate the CH4 and CO2 fluxes across the water-gas interface and identify their controlling factors, four diel field campaigns and one monthly sampling campaign during June 2010-May 2011 were carried out at a site near the Three Gorges Dam, China. The averaged CH4 and CO2 fluxes across the air-water interface from the site were much less than those reported from reservoirs in tropic and temperate regions, and from the natural river channels of the Yangtze River. CH4 Fluxes at the site were very low compared to most other reservoirs or natural lakes. One of the most important reasons may be due to the oxidation of CH4 in the water column owing to the great water depth and high DO in water in the Three Gorges Reservoir. The averaged monthly CH4 and CO2 fluxes at the site during the observation year were 0.05 mg/(m2 x hr) and 104.43 mg/(m2 x hr) respectively with the maximum occurred in July 2010. The monthly CO2 fluxes during the observation year were positively correlated to the surface water temperature, and negatively correlated to the air pressure and the surface water pH. The CO2 flux showed a positive correlation with DOC to some extent, although not significantly, which indicated that allochthonous organic C was a major source of CO2 and biogeochemical processes in this reservoir were C-limited. The significantly positive correlation between the reservoir outflow and the seasonal gas flux indicate the disturbance condition of the water body dominated the seasonal gas emission.