EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Study on determination of caffeic acid, chlorogenic acid in rat plasma and their pharmacokinetics with LC-MS/MS].
PMID 24494567
Abstract
To establish a LC-MS/MS method to determine caffeic acid, chlorogenic acid in rat plasma and study their pharmacokinetics in rats. Six Sprague-Dawley rats were intravenously injected with 4 mL x kg(-1) of Dengzhanxixin injection, respectively. Their drug plasma concentration was determined by LC-MS/MS, with tinidazole as an internal standard. The pharmacokinetic parameters were calculated by DAS 1.0. The linear concentration ranges of caffeic acid, and chlorogenic acid were 2-128 microg x L(-1) (r = 0.998 1) and 3-384 microg x L(-1) (r = 0.998 7), respectively. The methodological test showed conformance to the requirements. The intraday and inter-day variable coefficients were both less than 10.0%, indicating that both of legitimate precise and accuracy were in conformity with the requirements of biological sample analysis. For caffeic acid, the pharmacokinetic parameter t1/2beta AUC0-t, and CL were (130.91 +/- 38.77) min, (4.89 +/- 0.96) mg x min x L(-1) and (0.12 +/- 0.02) L x min(-1) x kg(-1), respectively. For chlorogenic acid, the pharmacokinetic parameter t1/2beta , AUC0-t, and CL were (49.38 +/- 8.85) min, (9.54 +/- 0.95) mg x min x L(-1) and (0.09 +/- 0.003) L x min(-1) x kg(-1), respectively. The LC-MS/MS analysis method established in this study was proved to be so accurate and sensitive that it can be applied to the pharmacokinetic study of caffeic acid and chlorogenic acid.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
51868
trans-Caffeic acid, certified reference material, TraceCERT®
C9H8O4