EMAIL THIS PAGE TO A FRIEND
Journal of internal medicine
Excessive activation of the alternative complement pathway in autosomal dominant polycystic kidney disease.
PMID 24494798
Abstract
The complement system is involved in many immune complex-mediated kidney diseases, yet its role in the pathogenesis of autosomal dominant polycystic kidney disease (ADPKD) has not been examined in detail. Screening of the glycoproteome of urine samples from ADPKD patients revealed that levels of complement factor B (CFB), serpin peptidase inhibitor, complement component 1 inhibitor (SERPING1) and complement component 9 (C9) increased, whereas complement component 1, r subcomponent-like (C1RL), CD55 and CD59 levels decreased with disease progression. Immunostaining and Western blot analysis confirmed the enhanced expression of CFB and C9 in cystic kidneys from ADPKD patients. Immunostaining also showed that the expressions of CFB and C9 in renal biopsy tissues from patients with other types of chronic kidney disease were lower than in tissues from ADPKD patients. The effect of the complement inhibitor rosmarinic acid (RMA) was evaluated in Pkd1(-/-) mice and Han:SPRD Cy/+ rats. Compared with vehicle-treated Pkd1(-/-) animals, RMA-treated mice had significantly lower serum creatinine (-50%) and blood urea nitrogen (-78%) levels, two kidneys/body weight ratio (-60%) and renal cystic index (-60%). Similar results were found in Cy/+ rats. Lower numbers of Ki67-positive nuclei and inflammatory cells and reduced fibrosis were observed in both animal models upon treatment with RMA. These results suggest that excessive activation of the alternative complement pathway is associated with ADPKD progression, probably mediated by cyst-lining epithelial cell proliferation, tubulointerstitial inflammatory cell infiltration and fibrosis. Targeting the complement system might represent a new therapeutic strategy for ADPKD.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1306373
ANTI-BF (CENTER) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA001817
Anti-CFB antibody produced in rabbit, Ab3, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA001832
ANTI-CFB antibody produced in rabbit, Ab4, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
H4759
Hydrochlorothiazide, crystalline
C7H8ClN3O4S2
H2910
Hydrochlorothiazide, meets USP testing specifications
C7H8ClN3O4S2
H1200000
Hydrochlorothiazide, European Pharmacopoeia (EP) Reference Standard
C7H8ClN3O4S2
1314009
Hydrochlorothiazide, United States Pharmacopeia (USP) Reference Standard
C7H8ClN3O4S2
PHR1032
Hydrochlorothiazide, Pharmaceutical Secondary Standard; Certified Reference Material
C7H8ClN3O4S2
Y0001494
Hydrochlorothiazide for peak identification, European Pharmacopoeia (EP) Reference Standard
C7H8ClN3O4S2
WH0004288M1
Monoclonal Anti-MKI67 antibody produced in mouse, clone 7B8, purified immunoglobulin, buffered aqueous solution
AMAB90870
Monoclonal Anti-MKI67 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1234, purified immunoglobulin, buffered aqueous glycerol solution
SAB5300423 Monoclonal Anti-MKI67 antibody produced in mouse, clone 4A1, ascites fluid
SAB5300424 Monoclonal Anti-MKI67 antibody produced in mouse, clone 9C12B2, ascites fluid
SAB5300425 Monoclonal Anti-MKI67 antibody produced in mouse, clone 8D5, ascites fluid
536954
Rosmarinic acid, 96%
C18H16O8
00390580
Rosmarinic acid, primary reference standard
C18H16O8
R4033
Rosmarinic acid, ≥98% (HPLC), from Rosemarinus officinalis L.
C18H16O8
Y0000786
Rosmarinic acid, European Pharmacopoeia (EP) Reference Standard
C18H16O8