EMAIL THIS PAGE TO A FRIEND
Voprosy pitaniia
[Pathogenetic bases of dietary prevention of dichlorobutene poisoning].
PMID 2449765
Abstract
It was shown experimentally that alimentary prevention of dichlorobutene intoxication could be realized in the presence of a protein-enriched ration, through additional injections of nutrients possessing antioxidant and lipotropic properties and playing a pathogenetically valid protective role in this intoxication.