EMAIL THIS PAGE TO A FRIEND
Blood
Quantitative phosphoproteomics unveils temporal dynamics of thrombin signaling in human endothelial cells.
PMID 24501219
Abstract
Thrombin is the key serine protease of the coagulation cascade and a potent trigger of protease-activated receptor 1 (PAR1)-mediated platelet aggregation. In recent years, PAR1 has become an appealing target for anticoagulant therapies. However, the inhibitors that have been developed so far increase bleeding risk in patients, likely because they interfere with endogenous PAR1 signaling in the endothelium. Because of its complexity, thrombin-induced signaling in endothelial cells has remained incompletely understood. Here, we have combined stable isotope amino acids in cell culture, affinity-based phosphopeptide enrichment, and high-resolution mass spectrometry and performed a time-resolved analysis of the thrombin-induced signaling in human primary endothelial cells. We identified 2224 thrombin-regulated phosphorylation sites, the majority of which have not been previously related to thrombin. Those sites were localized on proteins that are novel to thrombin signaling, but also on well-known players such as PAR1, Rho-associated kinase 2, phospholipase C, and proteins related to actin cytoskeleton, cell-cell junctions, and Weibel-Palade body release. Our study provides a unique resource of phosphoproteins and phosphorylation sites that may generate novel insights into an intimate understanding of thrombin-mediated PAR signaling and the development of improved PAR1 antagonists that affect platelet but not endothelial cell function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T4648 Thrombin from bovine plasma, lyophilized powder, 40-300 NIH units/mg protein (biuret)
T7513 Thrombin from bovine plasma, lyophilized powder, ≥2,000 NIH units/mg protein (E1%/280 = 19.5)
T6634 Thrombin from bovine plasma, lyophilized powder, 600-2,000 NIH units/mg protein (biuret)
T9549 Thrombin from bovine plasma, powder, cell culture tested, ≥1,500 NIH units/mg protein (E1%/280 = 19.5)
T7201 Thrombin from bovine plasma, lyophilized powder, ≥2,000 NIH units/mg protein (E1%/280 = 19.5)
T7326 Thrombin from bovine plasma, ≥60 NIH units/mg protein (biuret)
T6884
Thrombin from human plasma, lyophilized powder, ≥2,000 NIH units/mg protein (E1%/280, 18.3)
T7009
Thrombin from human plasma, lyophilized powder, ≥1,000 NIH units/mg protein (E1%/280, 18.3)
T1063
Thrombin from human plasma, lyophilized powder, ≥2800 NIH units/mg protein (E1%/280, 18.3)
T8885
Thrombin from human plasma, lyophilized powder, Suitable for routine use in the thrombin time test
T4393
Thrombin from human plasma, lyophilized powder, 1500-3500 NIH units/mg protein (E1%/280, 18.3), cell culture tested
T7572
Thrombin from human plasma, 400-1000 NIH units/mg protein
T5772 Thrombin from rat plasma, lyophilized powder, ≥1,000 units/mg protein (biuret)
T9326
Thrombin human, BioUltra, recombinant, expressed in HEK 293 cells, aqueous solution, ≥95% (SDS-PAGE)