EMAIL THIS PAGE TO A FRIEND
Klinichna khirurhiia
[Impact of saturation of the mixed capillary blood on the subjective estimation of the postoperative pain intensity].
PMID 24501968
Abstract
The impact of prolonged perineural blocade of n. femoralis on a subjective estimation of the postoperative pain intensity and its connection with the mixed capillary blood saturation was investigated.