EMAIL THIS PAGE TO A FRIEND
Klinichna Khirurhiia / Ministerstvo Okhorony zdorov'ia Ukrainy, Naukove Tovarystvo Khirurhiv Ukrainy
[Postoperative pancreonecrosis prophylaxis in a gastric cancer surgery].
PMID 24502000