EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
Biosynthesis of N,N-dimethyltryptamine (DMT) in a melanoma cell line and its metabolization by peroxidases.
PMID 24508833
Abstract
Tryptophan (TRP) is essential for many physiological processes, and its metabolism changes in some diseases such as infection and cancer. The most studied aspects of TRP metabolism are the kynurenine and serotonin pathways. A minor metabolic route, tryptamine and N,N-dimethyltryptamine (DMT) biosynthesis, has received far less attention, probably because of the very low amounts of these compounds detected only in some tissues, which has led them to be collectively considered as trace amines. In a previous study, we showed a metabolic interrelationship for TRP in melanoma cell lines. Here, we identified DMT and N,N-dimethyl-N-formyl-kynuramine (DMFK) in the supernatant of cultured SK-Mel-147 cells. Furthermore, when we added DMT to the cell culture, we found hydroxy-DMT (OH-DMT) and indole acetic acid (IAA) in the cell supernatant at 24 h. We found that SK-Mel-147 cells expressed mRNA for myeloperoxidase (MPO) and also had peroxidase activity. We further found that DMT oxidation was catalyzed by peroxidases. DMT oxidation by horseradish peroxidase, H2O2 and MPO from PMA-activated neutrophils produced DMFK, N,N-dimethyl-kynuramine (DMK) and OH-DMT. Oxidation of DMT by peroxidases apparently uses the common peroxidase cycle involving the native enzyme, compound I and compound II. In conclusion, this study describes a possible alternative metabolic pathway for DMT involving peroxidases that has not previously been described in humans and identifies DMT and metabolites in a melanoma cell line. The extension of these findings to other cell types and the biological effects of DMT and its metabolites on cell proliferation and function are key questions for future studies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L2005 Lactoperoxidase from bovine milk, lyophilized powder (essentially salt-free), ≥200 units/mg protein
61328 Lactoperoxidase from bovine milk, lyophilized, powder, ≥150 U/mg
M6908 Myeloperoxidase from human leukocytes, lyophilized powder, ≥50 units/mg protein
SML0791
N,N-Dimethyltryptamine, ≥97% (HPLC)
C12H16N2
68528 Peroxidase from Bjerkandera adusta, ≥2.8 U/mg
P8375 Peroxidase from horseradish, Type VI, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol)
77332 Peroxidase from horseradish, lyophilized, powder, beige, ~150 U/mg
P6782 Peroxidase from horseradish, Type VI-A, essentially salt-free, lyophilized powder, 950-2000 units/mg solid (using ABTS), ≥250 units/mg solid (using pyrogallol)
P8250 Peroxidase from horseradish, Type II, essentially salt-free, lyophilized powder, 150-250 units/mg solid (using pyrogallol)
P8125 Peroxidase from horseradish, Type I, essentially salt-free, lyophilized powder, 50-150 units/mg solid (using pyrogallol)
P8415 Peroxidase from horseradish, Type XII, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol)
P6140 Peroxidase from horseradish, Type X, ammonium sulfate suspension
P2088 Peroxidase from horseradish, Highly stabilized, essentially salt-free, lyophilized powder, 200-300 units/mg solid (using pyrogallol)