EMAIL THIS PAGE TO A FRIEND
Okajimas folia anatomica Japonica
Effects of the metal salts on the staining of elastic fibers with hematein.
PMID 2451196
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
51230
Hematein, for microscopy (Hist.)
C16H12O6