EMAIL THIS PAGE TO A FRIEND
Journal of clinical oncology : official journal of the American Society of Clinical Oncology
CD49d is the strongest flow cytometry-based predictor of overall survival in chronic lymphocytic leukemia.
PMID 24516016
Abstract
Although CD49d is an unfavorable prognostic marker in chronic lymphocytic leukemia (CLL), definitive validation evidence is lacking. A worldwide multicenter analysis was performed using published and unpublished CLL series to evaluate the impact of CD49d as an overall (OS) and treatment-free survival (TFS) predictor. A training/validation strategy was chosen to find the optimal CD49d cutoff. The hazard ratio (HR) for death and treatment imposed by CD49d was estimated by pooled analysis of 2,972 CLLs; Cox analysis stratified by center and stage was used to adjust for confounding variables. The importance of CD49d over other flow cytometry-based prognosticators (eg, CD38, ZAP-70) was ranked by recursive partitioning. Patients with ≥ 30% of neoplastic cells expressing CD49d were considered CD49d+. Decrease in OS at 5 and 10 years among CD49d+ patients was 7% and 23% (decrease in TFS, 26% and 25%, respectively). Pooled HR of CD49d for OS was 2.5 (2.3 for TFS) in univariate analysis. This HR remained significant and of similar magnitude (HR, 2.0) in a Cox model adjusted for clinical and biologic prognosticators. Hierarchic trees including all patients or restricted to those with early-stage disease or those age ≤ 65 years always selected CD49d as the most important flow cytometry-based biomarker, with negligible additional prognostic information added by CD38 or ZAP-70. Consistently, by bivariate analysis, CD49d reliably identified patient subsets with poorer outcome independent of CD38 and ZAP-70. In this analysis of approximately 3,000 patients, CD49d emerged as the strongest flow cytometry-based predictor of OS and TFS in CLL.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N9879 NADase from porcine brain, ≥0.007 unit/mg solid