EMAIL THIS PAGE TO A FRIEND
Journal of clinical oncology : official journal of the American Society of Clinical Oncology
Randomized phase III trial exploring the use of long-acting release octreotide in the prevention of chemotherapy-induced diarrhea in patients with colorectal cancer: the LARCID trial.
PMID 24516038
Abstract
Chemotherapy-induced diarrhea (CID) is a relatively common adverse event in the treatment of patients with colorectal cancer. The LAR for Chemotherapy-Induced Diarrhea (LARCID) trial evaluated the efficacy and safety of long-acting release octreotide (octreotide LAR) for the prevention of CID in this population. Patients with colorectal cancer starting adjuvant or first-line treatment with a chemotherapy combination containing fluorouracil, capecitabine, and/or irinotecan were randomly assigned to receive octreotide LAR 30 mg intramuscularly every 4 weeks (experimental arm) or the physician's treatment of choice in case of diarrhea (control arm). A total of 139 patients were randomly assigned, most of whom received fluorouracil- and oxaliplatin-containing chemotherapy regimens. The rate of diarrhea was 76.1% in the experimental group (n = 68) and 78.9% in the control group (n = 71). Treatment with octreotide LAR did not prevent or reduce the severity of CID. Treatment choices for diarrhea management included loperamide in the majority of patients. No benefit from octreotide LAR was identified in terms of need for diarrhea treatment, opioids, or intravenous hydration or in the rate of hospitalization or quality of life. This study could not prove the efficacy of octreotide LAR in the prevention of CID.