EMAIL THIS PAGE TO A FRIEND
Pakistan journal of biological sciences: PJBS
Differential role of ethylene and hydrogen peroxide in dark-induced stomatal closure.
PMID 24517017
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
223506
Calcium chloride dihydrate, ACS reagent, ≥99%
CaCl2 · 2H2O
C5080
Calcium chloride dihydrate, BioXtra, ≥99.0%
CaCl2 · 2H2O
C3881
Calcium chloride dihydrate, ReagentPlus®, ≥99.0%
CaCl2 · 2H2O
C7902
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
CaCl2 · 2H2O
C3306
Calcium chloride dihydrate, for molecular biology, ≥99.0%
CaCl2 · 2H2O
21097
Calcium chloride dihydrate, BioUltra, for molecular biology, ≥99.5% (KT)
CaCl2 · 2H2O
21098
Calcium chloride dihydrate, BioUltra, ≥99.5% (KT)
CaCl2 · 2H2O
31307
Calcium chloride dihydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
CaCl2 · 2H2O
12022
Calcium chloride dihydrate, puriss., meets analytical specification of Ph. Eur., USP, FCC, E509, 99-103%, ≤0.0001% Al
CaCl2 · 2H2O
21102
Calcium chloride dihydrate, tested according to Ph.Eur.
CaCl2 · 2H2O
H3410
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
H2O2
216763
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
H2O2
516813
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
H2O2
349887
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
H2O2
323381
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
H2O2
95299
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
H2O2
31642
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
H2O2
261130
Lanthanum, ingot (under oil), 99.9% trace rare earth metals basis
La
263117
Lanthanum, pieces
La
263109
Lanthanum, powder, −40 mesh, under oil, 99.9% trace rare earth metals basis
La
449830
Lanthanum(III) chloride, anhydrous, beads, −10 mesh, ≥99.99% trace metals basis
Cl3La
298182
Lanthanum(III) chloride, anhydrous, beads, −10 mesh, 99.9% trace metals basis
Cl3La
203521
Lanthanum(III) chloride heptahydrate, 99.999% trace metals basis
Cl3La · 7H2O
262072
Lanthanum(III) chloride heptahydrate, ACS reagent
Cl3La · 7H2O
31820
Lanthanum(III) chloride heptahydrate, puriss. p.a., ≥99%
Cl3La · 7H2O
P3130
Propyl gallate, powder
C10H12O5
02370
Propyl gallate, for microscopy, ≥98.0% (HPLC)
C10H12O5
48710
Propyl gallate, antioxidant, ≥98.0% (HPLC)
C10H12O5
W294705
Propyl gallate, ≥98%, FCC
C10H12O5
P3640000
Propyl gallate, European Pharmacopoeia (EP) Reference Standard
C10H12O5