EMAIL THIS PAGE TO A FRIEND
Microvascular Research
Increases in reactive oxygen species enhance vascular endothelial cell migration through a mechanism dependent on the transient receptor potential melastatin 4 ion channel.
PMID 24518820