EMAIL THIS PAGE TO A FRIEND
Journal of environmental sciences (China)
Cr(VI) removal from aqueous solution with bamboo charcoal chemically modified by iron and cobalt with the assistance of microwave.
PMID 24520714